Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Hydroxide Solution, MP Biomedical

Catalog No. MP215210450 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215210450 50 mL
MP215210425 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP215210450 Supplier MP Biomedicals, Inc Supplier No. 0215210450
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrabutylammonium Hydroxide Solution has a concentration of 40% in H2O and is a phase transfer catalyst.

  • Used as a curing accelerator for epoxy resins
  • Also used in preparation of dichlorocarbene from chloroform
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2052-49-5
Molecular Formula C16H37NO
Molecular Weight (g/mol) 259.48
MDL Number MFCD00009425
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1, tetrabutylammonium hydroxide solution in water
PubChem CID 2723671
IUPAC Name tetrabutylazanium hydroxide
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Physical Form Liquid
Quantity 50 mL
Packaging Glass
Density 0.99 g/mL at 25 °C (Lit.)
Grade Research
Concentration or Composition (by Analyte or Components) 40% in H2O
Chemical Name or Material Tetrabutylammonium Hydroxide
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.