Learn More
AdipoGen Tetracaine
SDP
Supplier:  AdipoGen CDXT0454G050
Encompass_Preferred
Chemical. CAS 94-24-6. Formula C15H24N2O. MW 264.36. Synthetic. Tetracaine is a potent local anaesthetic and a channel function allosteric inhibitor that blocks voltage-sensitive release of Ca2 from sarcoplasmic reticulum. At low concentrations, tetracaine causes an initial inhibition of spontaneous calcium release events, while at high concentrations, tetracaine blocks release completely. inhibitor of voltage-gated sodium channels such as NaV1.4 and NaV1.8. Shown also to reversibly block nicotinic acetylcholine ACh receptors nAChRs.
Catalog No. 50-255-9951
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.