Learn More
Description
Tetracaine USP is a white or light yellow, waxy solid. It is very slightly soluble in water and soluble in alcohol.
Alternative name(s): Tetracaine (Amethocaine)
Specifications
Specifications
| Chemical Name or Material | Tetracaine, USP |
| Packaging | Pail, 6 gallon, White, HDPE, Round |
| CAS | 94-24-6 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 264.369 |
| Quantity | 5 kg |
| Percent Purity | 98 to 102 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
