Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachloro-p-benzoquinone, 99%

Catalog No. AC213562500
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC213562500 250 g Plastic bottle
AC213561000 100 g Plastic bottle
AC213565000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC213562500 Supplier Thermo Scientific Chemicals Supplier No. 213562500
Add to Cart
This item is not returnable. View return policy

CAS: 118-75-2 | C6Cl4O2 | 245.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-75-2
Molecular Formula C6Cl4O2
Molecular Weight (g/mol) 245.86
MDL Number MFCD00001594
InChI Key UGNWTBMOAKPKBL-UHFFFAOYSA-N
Synonym chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone
PubChem CID 8371
ChEBI CHEBI:36703
IUPAC Name 2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione
SMILES ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O

Specifications

Melting Point >290°C
Flash Point >100°C
Loss on Drying 0.5% max. (1 g, 105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula C6Cl4(=O)2
Quantity 250 g
Beilstein 07, 636
Fieser 01,125; 02,66; 03,46; 04,75
Merck Index 15, 2078
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether,sparingly soluble in ccl4,cs2 and chloroform,practically insoluble in cold petroleum ether and,cold alcohol
Formula Weight 245.88
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetrachloro-p-benzoquinone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-274-4
RTECSNumber DK6825000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.