Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachloroauric(III) acid trihydrate, ACS reagent

Catalog No. AC411070250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411070250 25 g Glass bottle
AC411070010 1 g Glass bottle
AC411070050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC411070250 Supplier Thermo Scientific Chemicals Supplier No. 411070250

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

missing translation for 'validMainframeMsg'

Tetrachloroauric(III) Acid Trihydrate, >-49.0% (Au), HAuCl4.3H2O, CAS Number-16961-25-4, Gold trichloride Acid Trihydrate, Chloroauric Acid Trihydrate, 1g, Orange, 393.83, ACS Reagent, GHS Signal Word: Danger | CAS: 16961-25-4 | AuCl4H | 339.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16961-25-4
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
InChI Key VDLSFRRYNGEBEJ-UHFFFAOYSA-K
IUPAC Name trichlorogold hydrochloride
SMILES Cl.Cl[Au](Cl)Cl

Specifications

Color Orange
Physical Form Glistening Crystalline Powder or Lumps
Quantity 25 g
Packaging Glass bottle
Linear Formula HAuCl4·3H2O
Merck Index 15,212
Synonym Chloroauric acid trihydrate, Gold trichloride acid trihydrate
Solubility Information Solubility in water: 150g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 393.83
Percent Purity ≥49.0% (Au)
Grade ACS Reagent
Chemical Name or Material Tetrachloroauric(III) acid trihydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.