Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachloroethylene, ≥99.5%, Solstice

Catalog No. 60044935 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
6 x 1 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-935 2.5 L Glass Bottle
60-044-934 1 L Glass Bottle
60-044-937 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-044-935 Supplier Solstice Supplier No. 162112.5L

≥99.5%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-18-4
Molecular Formula C2Cl4
Molecular Weight (g/mol) 165.82
MDL Number MFCD00000834
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
PubChem CID 31373
ChEBI CHEBI:17300
IUPAC Name tetrachloroethene
SMILES ClC(Cl)=C(Cl)Cl

Specifications

Melting Point -22°C
Color Colorless
Density 1.623g/mL at 25°C
Boiling Point 121°C
Packaging Glass Bottle
Refractive Index n20/D 1.505(lit.)
Linear Formula CCl2=CCl2
Quantity 2.5 L
UN Number UN1897
Beilstein 1361721
Vapor Density 5.83 (vs air)
Vapor Pressure 13mmHg (20°C), 19mmHg (25°C)
Formula Weight 165.83g/mol
Percent Purity ≥99.5%
Physical Form Liquid
Chemical Name or Material Tetrachloroethylene
Show More Show Less
Hazard Statement P280-P305 + P351 + P338
Precautionary Statement H315-H317-H319-H336-H351-H411
DOTInformation 6.1/PGIII
EINECSNumber 204-825-9
RTECSNumber KX3850000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.