Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachlorofluorescein, MP Biomedicals

Catalog No. ICN219233 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN219233 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN219233 Supplier MP Biomedicals, Inc Supplier No. 0521923305

Chemical Identifiers

CAS 6262-21-1
Molecular Formula C20H8Cl4O5
Molecular Weight (g/mol) 470.08
MDL Number MFCD00010030
InChI Key QCPFFGGFHNZBEP-UHFFFAOYSA-N
PubChem CID 80424
IUPAC Name 4,5,6,7-tetrachloro-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
SMILES OC1=CC=C2C(OC3=CC(O)=CC=C3C22OC(=O)C3=C(Cl)C(Cl)=C(Cl)C(Cl)=C23)=C1

Specifications

Quantity 10 g
Chemical Name or Material Tetrachlorofluorescein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.