Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Tetracycline Hydrochloride, Broad Spectrum Antibiotic, Solid, 5 g

Catalog No. 502706786
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6786 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6786 Supplier Abcam Supplier No. AB1412235G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Broad-spectrum protein synthesis inhibitor with bacteriostatic activity.

  • Molecular Weight: 480.9 Da
  • Purity: >96%
  • Form: Solid
  • Properties: Broad spectrum antibiotic with bacteriostatic activity
  • Mechanism of Action: Protein synthesis inhibitor; binds to the 30S ribosomal subunit
  • Biological Effects: Antiapoptotic and anti-inflammatory effects; active in vivo
  • Composition: 100% Tetracycline hydrochloride

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at -20°C.
Shipping Condition Ambient
Buffer 100% Tetracycline hydrochloride
Form Solid
Target Molecule 30S Ribosomal Subunit
Formulation 100% Tetracycline hydrochloride
Quantity 5 g
For Use With (Application) Protein synthesis inhibition, anti-apoptotic research, anti-inflammatory research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.