Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Tetracycline hydrochloride, can be used as secondary standard

Catalog No. p-196510
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC233105000 500 mg Glass Bottle
AC233100025 2.5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC233105000 Supplier Thermo Scientific Chemicals Supplier No. 233105000
Only null left
Add to Cart

Tetracycline hydrochloride, CAS # 64-75-5, is a wide-spectrum antibiotic that targets both gram-negative and gram-positive bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Tetracycline hydrochloride is the hydrochloride salt of the antibiotic tetracycline
  • Tetracycline blocks amino acyl-tRNA binding during bacterial protein synthesis
  • Tetracycline is a broad-spectrum naphthacene antibiotic that inhibits growth of both gram positive and gram negative bacteria

Applications

  • Tetracycline may be used as a selection agent in cell culture
  • It may also be used to activate inducible gene expression in engineered cells
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-75-5
Molecular Formula C22H25ClN2O8
Molecular Weight (g/mol) 480.898
MDL Number MFCD00078142
InChI Key HTXDZWDXSWLLLW-FMZCEJRJSA-N
PubChem CID 129628373
IUPAC Name (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl

Specifications

Type Tetracycline Hydrochloride
Physical Form Powder
Melting Point 223.0°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation − 252.00
Color Yellow
Quantity 500 mg
Beilstein 14, IV, 2625
Merck Index 15, 9341
Solubility Information Solubility in water: 20g/L. Other solubilities: soluble in ethanol: 5g/L, insoluble in ether, hydrocarbons and acetone
Formula Weight 480.9
Percent Purity ≥95 (anhydrous and solvent free), 80 wt% (as is, calculated)
Chemical Name or Material Tetracycline hydrochloride
Show More Show Less
Hazard Category LIGHT SENSITIVE
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-593-8
RTECSNumber QI9100000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.