CAS RN 64-75-5
Tetracycline Hydrochloride, Fisher BioReagents
CAS: 64-75-5 Molecular Formula: C22H25ClN2O8 Molecular Weight (g/mol): 480.898 InChI Key: HTXDZWDXSWLLLW-FMZCEJRJSA-N PubChem CID: 129628373 IUPAC Name: (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride SMILES: CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl
Tetracycline hydrochloride, 96%
CAS: 64-75-5 Molecular Formula: C22H25ClN2O8 Molecular Weight (g/mol): 480.898 MDL Number: MFCD00078142 InChI Key: HTXDZWDXSWLLLW-FMZCEJRJSA-N PubChem CID: 129628373 IUPAC Name: (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride SMILES: CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl
Thermo Scientific Chemicals Tetracycline hydrochloride, can be used as secondary standard
CAS: 64-75-5 Molecular Formula: C22H25ClN2O8 Molecular Weight (g/mol): 480.898 MDL Number: MFCD00078142 InChI Key: HTXDZWDXSWLLLW-FMZCEJRJSA-N PubChem CID: 129628373 IUPAC Name: (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride SMILES: CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl
Tetracyline hydrochloride, MP Biomedicals™
CAS: 64-75-5 Molecular Formula: C22H25ClN2O8 Molecular Weight (g/mol): 480.898 InChI Key: HTXDZWDXSWLLLW-FMZCEJRJSA-N Synonym: tetracycline hydrochloride PubChem CID: 129628373 IUPAC Name: (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride SMILES: CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl
Tetracycline Hydrochloride 98.0+%, TCI America™
CAS: 64-75-5 Molecular Formula: C22H25ClN2O8 Molecular Weight (g/mol): 480.898 MDL Number: MFCD00078142 InChI Key: HTXDZWDXSWLLLW-FMZCEJRJSA-N Synonym: tetracycline hydrochloride PubChem CID: 129628373 IUPAC Name: (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride SMILES: CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl