Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracycline Hydrochloride USP , Medisca™
GREENER_CHOICESDP

Catalog No. 18048123 Shop All Medisca Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-048-123 25 g
18-048-044 100 g
18-048-062 500 g
18-048-063 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-048-123 Supplier Medisca Supplier No. 005304
Get 1 Extra Chemical When You Buy 4
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetracycline Hydrochloride USP is a yellow odorless crystalline powder. It is moderately hygroscopic. Stable in air, but exposure to strong sunlight in moist air causes it to darken.

Chemical Identifiers

CAS 64-75-5

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystalline Powder
Color Yellow
Quantity 25 g
Percent Purity ≥ 900 μg/mg (C22H24N2O8 HCl)
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.