Learn More
Gemini Bio Products GeminiBio Tetracycline Negative Fetal Bovine Serum, Low Tetracycline Levels, Sterile-Filtered, 500 mL
SDP
Supplier:  Gemini Bio Products 100800500
Encompass_Preferred
GeminiBio Tetracycline Negative Fetal Bovine Serum is specifically designed for sensitive cell culture applications, providing essential growth factors while minimizing tetracycline interference. It is triple-filtered for purity, mycoplasma-tested, and virus-screened, ensuring safe and reliable results. This serum allows for accurate gene expression control, making it ideal for research requiring high-quality materials. Store frozen at -20 to -10°C for optimal preservation.
Catalog No. NC1271788
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.