Learn More
Medchemexpress LLC Tetradec-13-ynoic acid | 82909-47-5 | MFCD18382104 | 95.0% | C14H24O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14033510MG
Encompass_Preferred
Alkynyl myristic acid (tetradec-13-ynoic acid) is an alkyne-terminated fatty acid used as a click-chemistry reagent and hydrophobic linker in chemical biology and medicinal chemistry. The terminal alkyne enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) to attach azide-functionalized probes or ligands, facilitating labeling, probe construction, and linker assembly for synthetic applications.
Catalog No. 50-002-60403