Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetradecafluorohexane 96.0+%, TCI America™
SDP

Catalog No. P0839250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0839250G 250 g
P083925G 25 g
P083910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0839250G Supplier TCI America Supplier No. P0839250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 355-42-0
Molecular Formula C6F14
Molecular Weight (g/mol) 338.044
MDL Number MFCD00000437
InChI Key ZJIJAJXFLBMLCK-UHFFFAOYSA-N
Synonym perfluorohexane, perflexane, tetradecafluorohexane, perfluoro-n-hexane, flutec pp1, hexane, tetradecafluoro, n-tetradecafluorohexane, fluorinert fc72, perfluorohexanes, perflexane usan:inn
PubChem CID 9639
ChEBI CHEBI:39427
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 60°C
Quantity 250 g
Formula Weight 338.04
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetradecafluorohexane
EINECSNumber (2)-2366
RTECSNumber MO4310000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.