Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetradecylcyclohexane 98.0+%, TCI America™
SDP

Catalog No. T087325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T087325G 25 g
T08735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T087325G Supplier TCI America Supplier No. T087325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1795-18-2
Molecular Formula C20H40
Molecular Weight (g/mol) 280.54
MDL Number MFCD00058951
InChI Key NQAVPKIJZCHUNS-UHFFFAOYSA-N
Synonym Myristylcyclohexane, 1-Cyclohexyltetradecane
PubChem CID 15715
IUPAC Name tetradecylcyclohexane
SMILES CCCCCCCCCCCCCCC1CCCCC1

Specifications

Color White
Quantity 25 g
Formula Weight 280.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Tetradecylcyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.