Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylammonium Hydroxide, 25% in Methanol, Thermo Scientific™

Catalog No. AC420301000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC420301000 100 g
AC420300250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC420301000 Supplier Thermo Scientific Chemicals Supplier No. 420301000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-98-5 , 67-56-1
Molecular Formula C8H21NO
Molecular Weight (g/mol) 147.26
MDL Number MFCD00009024
InChI Key LRGJRHZIDJQFCL-UHFFFAOYSA-M
Synonym tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
PubChem CID 6509
IUPAC Name tetraethylazanium hydroxide
SMILES [OH-].CC[N+](CC)(CC)CC

Specifications

Color Yellow
Linear Formula (C2H5)4NOH
Merck Index 15, 9346
Density 0.8500g/mL
Formula Weight 147.26
Percent Purity 24 to 26%
Quantity 100 g
Physical Form Solution
Chemical Name or Material Tetraethylammonium hydroxide, 25% in Methanol
Hazard Category Danger
EINECSNumber 201-073-3
RTECSNumber KH3150000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.