Learn More
Sigma Organic Chemistry Tetraethylene glycol | 1KG | 112-60-7 | MFCD00002879 | 0.99
Supplier: Sigma Organic Chemistry 1101751KG

Tetraethylene glycol, 1KG
About This Item:
Synonym(s): Bis[2-(2-hydroxyethoxy)ethyl] ether; Tetra(ethylene glycol); Tetraglycol
Linear Formula: HO(CH2CH2O)3CH2CH2OH
Molecular Weight: 194.23
CAS Number: 112-60-7
MDL Number: MFCD00002879
UNSPSC Code: 12162002
Purity: 0.99
Boiling Point: 314 C (lit.)
Melting Point: -5.6 C (lit.)
Density: 1.125 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.459 (lit.)
Storage: room temp
EINECS Number: 203-989-9
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.