Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrafluoroterephthalonitrile 98.0+%, TCI America™
SDP

Catalog No. T105025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T105025G 25 g
T10501G 1 g
T10505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T105025G Supplier TCI America Supplier No. T105025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1835-49-0
Molecular Formula C8F4N2
Molecular Weight (g/mol) 200.096
MDL Number MFCD00001776
InChI Key PCRSJGWFEMHHEW-UHFFFAOYSA-N
Synonym tetrafluoroterephthalonitrile, 2,3,5,6-tetrafluoroterephthalonitrile, diamond 2031, 2,3,5,6-tetrafluoro-1,4-benzenedicarbonitrile, 1,4-benzenedicarbonitrile, 2,3,5,6-tetrafluoro, 1,4-dicyanotetrafluorobenzene, terephthalonitrile, tetrafluoro, perfluoroterephthalonitrile, tetrafluoroterephthalodinitrile, tetrafluorobenzene-1,4-dicarbonitrile
PubChem CID 15783
IUPAC Name 2,3,5,6-tetrafluorobenzene-1,4-dicarbonitrile
SMILES C(#N)C1=C(C(=C(C(=C1F)F)C#N)F)F

Specifications

Melting Point 198°C
Color White-Yellow
Quantity 25 g
UN Number 3439
Formula Weight 200.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrafluoroterephthalonitrile
RTECSNumber CZ1955000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.