Learn More
Tetrahydrofuran, 99.8%, for HPLC, unstabilized
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.
Specifications
Specifications
| Absorbance | (1 cm cell vs water), 0.02 max. at 280nm, 0.04 max. at 270nm, 0.05 max. at 260nm, 0.1 max. at 250nm, 0.26 max. at 240nm, 0.5 max. at 230nm |
| Acidity | 0.0002 meq/g max. |
| Alkalinity | 0.0006 meq/g max. |
| Density | 0.8800g/mL |
| Quantity | 500 mL |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Residue after Evaporation | 0.0002% max. |
| Assay Percent Range | 99.8% |
| Vapor Pressure | Vapor Pressure: 200mbar at 20°C |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides. |
| Precautionary Statement | GHS P Statement: Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Avoid breathing dust/fume/gas/mist/vapors/spray. Avoid release to the environment. Keep container tightly closed. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.