Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, Puriss. p.a., contains 250 ppm BHT as inhibitor, ACS Reagent, Reag. Ph. Eur., ≥99.9%, Solstice

Catalog No. 60047707 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
6 x 1 L
175 kg
Packaging:
Aluminum Bottle
Glass Bottle
Metal Drum
N/A
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-707 6 x 1 L Glass Bottle
60-047-702 1 L Glass Bottle
60-047-703 2.5 L Glass Bottle
60-047-705 4 x 2.5 L N/A
60-047-701 175 kg Metal Drum
60-047-706 5 L Aluminum Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 60-047-707 Supplier Solstice Supplier No. 873686X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., contains 250 ppm BHT as inhibitor, ACS Reagent, Reag. Ph. Eur.,≥99.9%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point -108°C
Color Colorless
Density 0.887g/mL at 20°C, 0.889g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 0.118
Flash Point −17°C (1.4°F)
Ignition Point 610°F
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 114mmHg (15°C), 143mmHg (20°C)
UN Number UN2056
Beilstein 102391
Boiling Point 65°C (1013 hPa)
Formula Weight 72.11g/mol
Percent Purity ≥99.9%
Chemical Name or Material Tetrahydrofuran
Show More Show Less
Hazard Statement P210-P280-P301 + P312 + P330-P305 + P351 + P338-P370 + P378-P403 + P235
DOTInformation 3/PGII
RTECSNumber 203-726-8
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.