Learn More
Sigma Aldrich Fine Chemicals Biosciences Tetrahydrofuran anhydrous
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 401757
Encompass
Tetrahydrofuran (THF) is a saturated cyclic ether with a potential use as a biofuel. Its combustion studies have been investigated. Reports suggest that it is a better promoter than 1,3 dioxolane for CO2-hydrate formation.
Catalog No. 50-290-7344
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.