Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, Anhydrous, 99.9% min., MilliporeSigma™

Catalog No. MTX02777 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MTX02777 100 mL
MTX02776 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MTX02777 Supplier MilliporeSigma Supplier No. US1TX0277/7
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point -108.3°C
Density 0.89g/cm3 at 20°C
Quantity 100 mL
Flash Point 15.15°C (4.7°F), Closed cup
Ignition Point 320.85°C (609.5°F)
Odor Ether-like
Physical Form Liquid
Assay Percent Range 99.9%, min.
Linear Formula CH2(CH2)3O
UN Number UN 2056
Boiling Point 66.1°C
Solubility Information Soluble in water
Formula Weight 72.11
Chemical Name or Material Tetrahydrofuran
Show More Show Less
DOTInformation DOT Class: 3, Packaging Group: II
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.