Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, BioSyn™, for biosynthesis and applications requiring low water content, contains BHT, >99.9%, Solstice

Catalog No. 60048595 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-595 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 60-048-595 Supplier Solstice Supplier No. BB3434
Add to Cart
Add to Cart
This item is not returnable. View return policy
  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

CAS Max % 100.00
Density 0.8892 g/cm3
Quantity 4 x 4 L
Flash Point -14°C
Packaging Glass Bottle
Physical Form Liquid
Boiling Point 66°C
Grade Low Water
Hazard Category Danger
Hazard Statement H351+H335+H319+H302+H225
Precautionary Statement P201+P202+P210+P233+P240+P241+P242+P243+P261+P264+P270+P271+P280
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.