Learn More
Sigma Aldrich Fine Chemicals Biosciences Tetrahydrofuran contains 2200L
Supplier: Sigma Aldrich Fine Chemicals Biosciences 360589
Tetrahydrofuran (THF) is a widely used organic solvent. On long term storage it forms organic peroxides. This process can be suppressed by adding butylated hydroxytoluene (BHT) as a stabilizer. BHT removes the free radicals required for the peroxide formation. The effect of THF on the biological systems has been studied.