Learn More
Tetrahydrofuran-D8, deuteration degree min 99.5% for NMR spectroscopy MagniSolv™, MilliporeSigma™
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Physical Form | Liquid |
| Packaging | Glass Ampule |
| Quantity | 10 x 0.75 mL |
| Color | Colorless |
| Odor | Ether-like |
| Assay Percent Range | 99.5% min. |
| Formula Weight | 80.16 |
| Chemical Name or Material | Tetrahydrofuran-D8 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.