Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, Solstice

Catalog No. 60090568 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
Returnable NOWPAK™ II
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-568 20 L Returnable NOWPAK™ II
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 60-090-568 Supplier Solstice Supplier No. BN34320
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BioSyn™, for biosynthesis and applications requiring low water content, contains BHT, >99.9%

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9
PubChem CID 8028
ChEBI CHEBI:26911

Specifications

Quantity 20 L
Flash Point -14 °C
Packaging Returnable NOWPAK™ II
Physical Form Liquid
Grade Low Water
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.