Learn More
Sigma Aldrich Fine Chemicals Biosciences Tetrahydrofuran inhibitor
Supplier: Sigma Aldrich Fine Chemicals Biosciences 34865
Tetrahydrofuran (THF) is widely employed as a solvent. It constitutes the key fragment of various natural products (polyether antibiotics). THF forms a double hydrate with hydrogen sulfide. Crystal structure of this double hydrate has been investigated by three-dimensional single-crystal studies. Butane-14-diol is formed as an intermediate during the synthesis of THF. Hot THF is useful for the dissolution of polyvinylidene chloride (PVDV).