Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, Stabilized with 0.025% BHT, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18614358 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
120 mL
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-358 100 mL Amber Glass Bottle
18-614-359 120 mL Amber Glass Bottle
18-614-360 500 mL Amber Glass Bottle
18-614-361 1 L Amber Glass Bottle
18-614-362 4 L Amber Glass Bottle
18-614-363 20 L Steel Can
18-614-364 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 18-614-358 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T1025100ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 109-99-9
Molecular Weight (g/mol) 72.11

Specifications

CAS Max % 1
Quantity 100 mL
Identification To Pass Test
Packaging Amber Glass Bottle
Residue after Evaporation 0.0003
Water 0.0005
Boiling Point 65°C to 66°C
Percent Purity 0.99
Grade Reagent
Chemical Name or Material Tetrahydrofuran
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.