Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, Stabilized, Technical, Spectrum™ Chemical
SDP

Catalog No. 18614355 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Amber Glass Bottle
Steel Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-355 500 mL Amber Glass Bottle
18-614-357 20 L Steel Can
18-614-356 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-614-355 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T1226500MLGL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 109-99-9
Molecular Weight (g/mol) 72.11

Specifications

CAS Max % 1
Quantity 500 mL
Packaging Amber Glass Bottle
Boiling Point 65°C to 66°C
Grade Technical
Chemical Name or Material Tetrahydrofuran
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.