Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran UltimAR, Macron Fine Chemicals™

Catalog No. MKV5584 Shop All Avantor Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-V558-4 4 L Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MK-V558-4 Supplier Avantor Supplier No. V55810
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Quantity 4 L
Packaging Glass
Formula Weight 72.11
Chemical Name or Material Tetrahydrofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.