Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofurfuryl Bromide 95.0+%, TCI America™
SDP

Catalog No. T1808100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1808100G 100 g
T180825G 25 g
T1808500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1808100G Supplier TCI America Supplier No. T1808100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1192-30-9
Molecular Formula C5H9BrO
Molecular Weight (g/mol) 165.03
MDL Number MFCD00016894
InChI Key VOHILFSOWRNVJJ-UHFFFAOYNA-N
Synonym tetrahydrofurfuryl bromide, 2-bromomethyl tetrahydrofuran, 2-bromomethyl oxolane, furan, 2-bromomethyl tetrahydro, 2-bromomethyl-tetrahydrofuran, furan, tetrahydro-2-bromomethyl, tetrahydrofurfurylbromide, tetrahydrofrufryl bromide, 2-bromomethyltetrahydrofuran
PubChem CID 14501
IUPAC Name 2-(bromomethyl)oxolane
SMILES BrCC1CCCO1

Specifications

Color Yellow
Quantity 100 g
Physical Form Clear Liquid at 20°C
Boiling Point 70°C
Formula Weight 165.03
Percent Purity ≥95.0% (GC)
Chemical Name or Material Tetrahydrofurfuryl Bromide
EINECSNumber (9)-0494
RTECSNumber LU6000000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.