Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofurfuryl Butyrate 98.0+%, TCI America™
SDP

Catalog No. T087825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T087825ML 25 mL
T0878500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T087825ML Supplier TCI America Supplier No. T087825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2217-33-6
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.224
MDL Number MFCD00059734
InChI Key DPZVDLFOAZNCBU-UHFFFAOYSA-N
Synonym Butyric Acid Tetrahydrofurfuryl Ester, 2-(Butyryloxymethyl)tetrahydrofuran
PubChem CID 551316
IUPAC Name oxolan-2-ylmethyl butanoate
SMILES CCCC(=O)OCC1CCCO1

Specifications

Color Yellow
Quantity 25 mL
Physical Form Clear Liquid at 20°C
Boiling Point 227°C
Formula Weight 172.22
Percent Purity ≥98.0% (GC)
Chemical Name or Material Tetrahydrofurfuryl Butyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.