Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofurfurylamine 98.0+%, TCI America™
SDP

Catalog No. T1072500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1072500ML 500 mL
T107225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1072500ML Supplier TCI America Supplier No. T1072500ML

Chemical Identifiers

CAS 4795-29-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00005373
InChI Key YNOGYQAEJGADFJ-UHFFFAOYNA-N
Synonym tetrahydrofurfurylamine, 2-aminomethyltetrahydrofuran, 2-tetrahydrofurfurylamine, tetrahydrofuran-2-yl methanamine, 2-furanmethanamine, tetrahydro, furfurylamine, tetrahydro, 2-aminomethyl tetrahydrofuran, usaf q-2, tetrahydro-2-furanmethanamine, oxolan-2-yl-methanamine
PubChem CID 253298
IUPAC Name 1-(oxolan-2-yl)methanamine
SMILES NCC1CCCO1

Specifications

Color Yellow
Quantity 500 mL
Physical Form Clear Liquid at 20°C
UN Number 2943
Boiling Point 155°C
Formula Weight 101.15
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material Tetrahydrofurfurylamine
EINECSNumber (9)-0854
RTECSNumber LV0175000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.