Learn More
Sigma Organic Chemistry Tetrahydropyran-2-ylmet | 50mg | 6628-83-7 | MFCD00129175
Supplier: Sigma Organic Chemistry CDS00645850MG

Tetrahydropyran-2-ylmet , 50mg
About this Item:
CAS #: 6628-83-7
MDL #: MFCD00129175
Molecular Weight: 115.17
UNSPSC Code: 12352200
Chemical Formula: C6H13NO
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.