Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydropyran-4-methanol 97.0+%, TCI America™
SDP

Catalog No. T29655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29655G 5 g
T29651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29655G Supplier TCI America Supplier No. T29655G

Chemical Identifiers

CAS 14774-37-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00457804
InChI Key YSNVSVCWTBLLRW-UHFFFAOYSA-N
Synonym tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol
PubChem CID 2773573
IUPAC Name (oxan-4-yl)methanol
SMILES OCC1CCOCC1

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 116.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetrahydropyran-4-methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.