Learn More
Sigma Organic Chemistry Tetrahydrothiopyran-4-o | 1G | 6309-59-7 | MFCD00102049
Supplier: Sigma Organic Chemistry SYX003641G

Tetrahydrothiopyran-4-o , 1G
About this item:
CAS #: 6309-59-7
MDL #: MFCD00102049
Chemical Formula: C5H9NOS
Molecular Weight: 131.2
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.