Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraiodo-2-sulfobenzoic Anhydride 95.0+%, TCI America™
SDP

Catalog No. T01311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T01311G 1 g
T013125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T01311G Supplier TCI America Supplier No. T01311G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1745-83-1
Molecular Formula C7I4O4S
Molecular Weight (g/mol) 687.751
MDL Number MFCD00059713
InChI Key UQDQTEHMZFWZSX-UHFFFAOYSA-N
PubChem CID 636253
IUPAC Name 4,5,6,7-tetraiodo-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-one
SMILES C12=C(C(=C(C(=C1I)I)I)I)S(=O)(=O)OC2=O

Specifications

Color Yellow
Quantity 1 g
UN Number 1759
Formula Weight 687.75
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraiodo-2-sulfobenzoic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.