Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(4-bromophenyl)ethylene 97.0+%, TCI America™
SDP

Catalog No. T34231G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34231G 1 g
T34235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34231G Supplier TCI America Supplier No. T34231G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61326-44-1
Molecular Formula C26H16Br4
Molecular Weight (g/mol) 648.03
MDL Number MFCD00667761
InChI Key BIRLDGKMJJEZRI-UHFFFAOYSA-N
PubChem CID 11828419
IUPAC Name 1-bromo-4-[1,2,2-tris(4-bromophenyl)ethenyl]benzene
SMILES BrC1=CC=C(C=C1)C(=C(C1=CC=C(Br)C=C1)C1=CC=C(Br)C=C1)C1=CC=C(Br)C=C1

Specifications

Melting Point 260°C
Color White
Quantity 1 g
Formula Weight 648.03
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrakis(4-bromophenyl)ethylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.