Learn More
Sigma Organic Chemistry Tetrakis(dimethylamido)titanium(IV) | 25G | 3275-24-9 | MFCD00014861 | 99.999% (trace metals analysis)
Supplier: Sigma Organic Chemistry 66900825G

Tetrakis(dimethylamido)titanium(IV), 25G
About This Item:
Storage: room temp
EINECS Number: 221-904-3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.