Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(dimethylamino)ethylene 98%, Thermo Scientific™

Catalog No. AC215300250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC215300250 25 g
AC215301000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC215300250 Supplier Thermo Scientific Chemicals Supplier No. 215300250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 996-70-3
Molecular Formula C10H24N4
Molecular Weight (g/mol) 200.33
InChI Key CBXRMKZFYQISIV-UHFFFAOYSA-N
Synonym tetrakis dimethylamino ethylene, ethenetetramine, octamethyl, tdae, unii-p6737b68mn, n1,n1,n'1,n'1,n2,n2,n'2,n'2-octamethylethene-1,1,2,2-tetraamine, octamethylethylenetetramine, 1,1,2,2-ethenetetramine, n1,n1,n1',n1',n2,n2,n2',n2'-octamethyl, dimethyl 1,2,2-tris dimethylamino vinyl amine, acmc-209sdr, tetra dimethlamino ethylene
PubChem CID 70455
IUPAC Name 1-N,1-N,1-N',1-N',2-N,2-N,2-N',2-N'-octamethylethene-1,1,2,2-tetramine
SMILES CN(C)C(=C(N(C)C)N(C)C)N(C)C

Specifications

Color Yellow
Density 0.86
Infrared Spectrum Authentic
Refractive Index 1.4790 to 1.4830 (20°C, 589nm)
Linear Formula [(CH3)2N]2C=C[N(CH3)2]2
Quantity 25 g
Beilstein 04, IV, 167
Merck Index 15, 9366
Formula Weight 200.33
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material Tetrakis(dimethylamino)ethylene, 98%
Show More Show Less
Hazard Category AIR SENSITIVE
EINECSNumber 213-638-1
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.