Learn More
MBL International Corporation H-2Kb MuLV p15E Tetramer-KSPWFTTL-PE
SDP
Supplier:  MBL International Corporation TBM5071
Encompass
Allele:H-2Kb; Application:FCM; Conjugate:PE; Peptide Sequence:KSPWFTTL; Species Reactivity:Mouse; Size:50 tests; Target:MuLV; MHC Tetramers are used for the detection of antigen-specific T-cell populations. MHC-peptide tetramer complexes recognize antigen specific T-cells at the single cell level, allowing for accurate measurements of the functional immune response in cancer, infectious diseases, and autoimmune diseases.  Due to their increased avidity, MHC tetramers stably bind to cognate TcRs and are evaluated using flow cytometry.
Catalog No. 50-168-9385
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.