Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethyl-1,3-cyclobutanedione 99.0+%, TCI America™
SDP

Catalog No. D025825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D025825G 25 g
D02585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D025825G Supplier TCI America Supplier No. D025825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 933-52-8
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.182
MDL Number MFCD00001331
InChI Key RGCDVHNITQEYPO-UHFFFAOYSA-N
Synonym tetramethyl-1,3-cyclobutanedione, tetramethylcyclobutane-1,3-dione, 1,3-cyclobutanedione, 2,2,4,4-tetramethyl, tetramethylcyclobuta-1,3-dione, 2,2,4,4-tetramethyl-1,3-cyclobutanedione, unii-rt4aq22ks4, 1,1,3,3-tetramethylcyclobutanedione, 2,2,4,4-tetramethylcyclobutanedione, dimethyl ketene dimer, rt4aq22ks4
PubChem CID 13617
IUPAC Name 2,2,4,4-tetramethylcyclobutane-1,3-dione
SMILES CC1(C(=O)C(C1=O)(C)C)C

Specifications

Melting Point 114°C
Color White-Yellow
Boiling Point 159°C
Quantity 25 g
Formula Weight 140.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetramethyl-1,3-cyclobutanedione [Precursor to Dimethyl Ketene]
RTECSNumber GU1775000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.