Learn More
Tetramethylammonium bromide, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Tetramethylammonium bromide acts as an ion pair reagent. It is used as a surface-active agent and an active ingredient for conditioners. Further, it is used as a softner for textiles and paper products. It is also employed as a disinfection agent, sanitizer, antimicrobial and antistatic agent. In addition to this, it plays in important role as a phase transfer catalyst in synthetic chemistry.
Solubility
Soluble in water, alcohol, chloroform and acetone.
Notes
Hygroscopic. Incompatible with oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | >300°C |
| Density | 1.56 |
| Odor | Odorless |
| Linear Formula | (CH3)4NBr |
| Quantity | 2500 g |
| UN Number | UN2811 |
| Beilstein | 3620955 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water,alcohol,chloroform and acetone. |
| Formula Weight | 154.06 |
| Show More |
Safety and Handling
| Hazard Category | H300-H315-H319-H335 |
| Hazard Statement | GHS H Statement H300-H315-H319-H335 Fatal if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 200-581-2 |
| RTECSNumber | BS7600000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.