Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium Hydroxide, 1% in Methanol, Reagents

Catalog No. 01333463
Change view
Click to view available options
:
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-333-463 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-463 Supplier Reagents Holdings Llc Supplier No. CT0077001C
Only null left
Add to Cart
Add to Cart

Tetramethylammonium Hydroxide, 1% in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 75-59-2 , 67-56-1
Molecular Formula C4H13NO
Molecular Weight (g/mol) 91.15
InChI Key WGTYBPLFGIVFAS-UHFFFAOYSA-M
IUPAC Name tetramethylazanium hydroxide
SMILES [OH-].C[N+](C)(C)C

Specifications

Color Colorless to Light Yellow
pH 12 to 13
Boiling Point 65°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 0.7943
Percent Purity 1%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Tetramethylammonium Hydroxide, 1% in Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.