Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium Sulfate 98.0+%, TCI America™
SDP

Catalog No. T124025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T124025G 25 g
T12405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T124025G Supplier TCI America Supplier No. T124025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14190-16-0
Molecular Formula C8H24N2O4S
Molecular Weight (g/mol) 244.35
MDL Number MFCD00012139
InChI Key KJFVITRRNTVAPC-UHFFFAOYSA-L
PubChem CID 14746849
IUPAC Name tetramethylazanium;sulfate
SMILES C[N+](C)(C)C.C[N+](C)(C)C.[O-]S(=O)(=O)[O-]

Specifications

Melting Point 285°C
Color White
Quantity 25 g
Formula Weight 244.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylammonium Sulfate
EINECSNumber (2)-0186
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.