Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Tetramethylrhodamine-5-maleimide, Labeling Reagent, Thiol Reactive, 1 mg

Catalog No. 502705189
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5189 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5189 Supplier Abcam Supplier No. AB1454431MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiol-reactive fluorescent probe for use in cytofluorescence applications.

  • Molecular Weight: 481.5 g/mol
  • Purity: >85%
  • Application: Thiol reactive fluorescent probe for use in cytofluorescence
  • Composition: 100% Tetramethylrhodamine-5-maleimide
  • CAS Number: 174568-67-3

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Proteins (Thiols)
Substrate Properties Chemical Substrate
Substrate Protein Thiols
Label or Dye Tetramethylrhodamine
Shipping Condition Ambient
Product Type Protein Labeling Reagent
Buffer 100% Tetramethylrhodamine-5-maleimide
Content And Storage Store at -20°C.
Detection Location In-Cell Detection
Detection Method Fluorescence
Formulation 100% Tetramethylrhodamine-5-maleimide
Quantity 1 mg
For Use With (Application) Cytofluorescence, protein labeling
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.