Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Tetramethylrhodamine-5-maleimide, Protein Thiol Labeling Reagent, >85% Purity, 25 mg

Catalog No. 502705188
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5188 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5188 Supplier Abcam Supplier No. AB14544325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiol reactive fluorescent probe for use in cytofluorescence.

  • Molecular weight: 481.5 g/mol
  • Purity: >85%
  • Application: Thiol-reactive fluorescent probe for use in cytofluorescence
  • Composition: 100% Tetramethylrhodamine-5-maleimide

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Proteins (Thiols)
Substrate Properties Chemical Substrate
Substrate Protein Thiols
Label or Dye Tetramethylrhodamine
Shipping Condition Ambient
Product Type Protein Labeling Reagent
Buffer 100% Tetramethylrhodamine-5-maleimide
Detection Location In-Cell Detection
Detection Method Fluorescence
Content And Storage Store at -20°C.
Formulation 100% Tetramethylrhodamine-5-maleimide
Quantity 25 mg
For Use With (Application) Protein thiol labeling, cytofluorescence
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.