Learn More
Abcam Tetramethylrhodamine-5-maleimide, Thiol Reactive Fluorescent Probe, >85% Purity, 5 mg
Description
- Thiol reactive fluorescent probe for use in cytofluorescence
- Molecular weight: 481.5 g/mol
- Purity: >85%
- Constituents: 100% Tetramethylrhodamine-5-maleimide
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Label or Dye | Tetramethylrhodamine |
| Reactivity | Protein thiols |
| Shipping Condition | Ambient |
| Substrate | Protein Thiols |
| Buffer | 100% Tetramethylrhodamine-5-maleimide |
| Detection Location | In-Cell Detection |
| Product Type | Protein Labeling Reagent |
| Content And Storage | Store at -20°C. |
| Target Molecule | Proteins (Thiols) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.