Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylsilane, 99%

Catalog No. ac268861000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC268861000 100 mL Glass bottle
AC268865000 500 mL Glass bottle
AC268860025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC268861000 Supplier Thermo Scientific Chemicals Supplier No. 268861000
Only null left
Add to Cart

CAS: 75-76-3 | C4H12Si | 88.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-76-3
Molecular Formula C4H12Si
Molecular Weight (g/mol) 88.23
MDL Number MFCD00008274
InChI Key CZDYPVPMEAXLPK-UHFFFAOYSA-N
Synonym silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073
PubChem CID 6396
ChEBI CHEBI:85361
IUPAC Name tetramethylsilane
SMILES C[Si](C)(C)C

Specifications

Melting Point -99.0°C
Color Colorless
Density 0.6400g/mL
Boiling Point 26.0°C to 28.0°C
Flash Point −27°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.3585
Linear Formula Si(CH3)4
Quantity 100 mL
Beilstein 04, 625
Specific Gravity 0.64
Merck Index 15, 9377
Solubility Information Solubility in water: 0.02g/L (25°C). Other solubilities: soluble in most common organic solvents
Formula Weight 88.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetramethylsilane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Extremely flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-899-1
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.