Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylthiuram disulfide, 97%

Catalog No. AAB200630E
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB200630E 2500 g
AAB2006322 100 g
AAB2006336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB200630E Supplier Thermo Scientific Chemicals Supplier No. B200630E
Add to Cart

CAS: 137-26-8 | C6H12N2S4 | 240.416 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-26-8
Molecular Formula C6H12N2S4
Molecular Weight (g/mol) 240.416
MDL Number MFCD00008325
InChI Key KUAZQDVKQLNFPE-UHFFFAOYSA-N
Synonym thiram, tetramethylthiuram disulfide, thiuram, tmtd, pomarsol, thirame, arasan, fernasan, nobecutan, rezifilm
PubChem CID 5455
ChEBI CHEBI:9495
IUPAC Name dimethylcarbamothioylsulfanyl N,N-dimethylcarbamodithioate
SMILES CN(C)C(=S)SSC(=S)N(C)C

Specifications

Melting Point 152°C to 155°C
Density 1.3
Boiling Point 129°C (20 mmHg)
Flash Point 89°C (192°F)
Quantity 2500 g
UN Number UN3077
Beilstein 1725821
Merck Index 14,9371
Formula Weight 240.43
Percent Purity 97%
Chemical Name or Material Tetramethylthiuram disulfide
Show More Show Less
Hazard Category H302-H315-H317-H319-H373-H500
Hazard Statement GHS H Statement
H373-H302-H332-H315-H319-H317
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P314-P330-P333+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 205-286-2
RTECSNumber JO1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.